
2-tert-Butylaniline 98%
ACRO369870250
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Appearance:Clear light yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5460 (20°C, 589 nm)
- GC:≥97.5 %
:
:149.24 g/mol
:123…124 °C (17 mmHg)
:–60 °C
:0.957 g/cm³
:102 °C
:228-634-5
:GHS07
:6310-21-0
:C₁₀H₁₅N
:MFCD00130023