Orders LinkContactUsLinkComponent
Screwthread tubes, attachable, DIN thread, DURAN®
Screwthread tubes, attachable, DIN thread, DURAN®
Supplier:   DWK Life Sciences
Screwthread tubes, attachable, DIN thread, DURAN®
Supplier:  DWK Life Sciences
Certificates
Screwthread tubes, attachable, DIN thread, DURAN®
Screwthread tubes, attachable, DIN thread, DURAN®
5 Options Available Below
Product Details & Documents
Made of DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
DIN 12216
Product Family Options
Product Information
  • Thread
    Tube ext.Ø
    Wall thickness
    Total length
    Availability
    Price
  • GL 14
    12 mm
    1,5 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 1 090,00 Kč
    1 090,00 Kč
    1 * 10 KS
       1 090,00 Kč
  • GL 18
    16 mm
    1,8 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 1 270,00 Kč
    1 270,00 Kč
    1 * 10 KS
       1 270,00 Kč
  • GL 25
    22 mm
    1,8 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 1 660,00 Kč
    1 660,00 Kč
    1 * 10 KS
       1 660,00 Kč
  • GL 32
    28 mm
    2,0 mm
    140 mm
    Log in for availability
    Log in for availability
    1 * 10 KS
    1 * 10 KS - 3 080,00 Kč
    3 080,00 Kč
    1 * 10 KS
       3 080,00 Kč
  • GL 45
    40 mm
    2,3 mm
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 1 010,00 Kč
    1 010,00 Kč
    1 * 1 KS
       1 010,00 Kč