α,α,α-Trifluoro-3-nitrotoluene
Katalogové číslo PROOMM0161.10
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:191.11 g/mol
Boiling Pt:202.8 °C (1013 hPa)
Melting Pt:–2.4 °C
Density:1.437 g/cm³ (20 °C)
EINECS:202-670-1
UN:2306
ADR:6.1,II
CAS Number:98-46-4
Formula:C₇H₄F₃NO₂
MDL Number:MFCD00007260