

2,6-Dichloro-3-nitrotoluene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29682-46-0
:
Formula:C₇H₅Cl₂NO₂
MDL Number:MFCD00024185
:206.03 g/mol
Melting Pt:53…55 °C
Storage Temperature:Ambient
EINECS:249-776-4
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together