
Acetophenone for synthesis
Catalog # 8.00028.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:5.5 g/l (20°C)
- Melting Point:20 °C
- Boiling Point:202 °C (1013 hPa)
- Vapor Pressure:0.4 hPa (20 °C)
- Flash Point:82 °C
- Refractive Index:1.5339 (20 °C, 589 nm)
- Explosion Limit:1.4 - 5.2 %(V)
- Density:1.03 g/cm3 (20 °C)
- Ignition Point:570°C
- Synonyms:Methyl phenyl ketone, 1-Phenylethanone, Phenylethanone, ACP
- Hill Formula:C8H8O
- Chemical Formula:CH3COC6H5
Specification Test Results
About this item
:
:120.15 g/mol
Merck Index:13,00074
Boiling Pt:201.7 °C (1013 hPa)
Melting Pt:20 °C
Density:1.027 g/cm³ (20 °C)
Flash Pt:82 °C
Storage Temperature:Ambient
EINECS:202-708-7
UN:3334
ADR:9,
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:98-86-2
Formula:C₈H₈O
MDL Number:MFCD00008724