Foot for rods
80 mm diameter, Rods (Ø 12 mm)
Catalog # BOCH5060
Supplier: BOCHEM
Jump to:
Product Details & Documents
Tempered cast iron, green varnished.
Specifications
- For:Rods (Ø 12 mm)
- Description:80 mm diameter
Tempered cast iron, green varnished.