(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, Supelco®
Catalog # 65361-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:234.17 g/mol
:M-Clarity™ quality level = MQ100
EINECS:243-829-5
CAS Number:20445-31-2
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD00004184