11-Chloro-1-undecene, Sigma-Aldrich®
Catalog # SIAL593117-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:188.74 g/mol
CAS Number:872-17-3
Formula:C₁₁H₂₁Cl
MDL Number:MFCD00671354