Isododecane (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:31807-55-3
:
Formula:C₁₂H₂₆
MDL Number:MFCD00036106
:170.34 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:250-816-8
UN:3295
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together