
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20445-31-2
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD00004184
:234.17 g/mol
Merck Index:14,6280
Boiling Pt:105…107 °C (1 mmHg)
Melting Pt:43…45 °C
Density:1.344
Flash Pt:>110 °C (230 °F)
EINECS:243-829-5
:GHS07
Related Information
Frequently Bought Together