Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Dodecane (mixture of isomers)
 
undefined
 
 
Dodecane (mixture of isomers)

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4190 to 1.4230 (20°C, 589 nm)
  • Density:
    (15°C) 746 to 756 kg/m3
  • Distillation range:
    170 to 195 °C
  • Aromatics:
    ≤50 ppm

 

 

Frequently Bought Together