
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20445-31-2
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD00004184
:234.17 g/mol
Boiling Pt:116…118 °C (1.5 mmHg)
Melting Pt:46…49 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:243-829-5
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid or low melting solid
- Infrared spectrum:Conforms
- Specific optical rotation:+71° to +74° (20°C, 589 nm) (c=2, CH3OH)
- Titration with NaOH:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together