
Phenethyl bromide 98%
:103-63-9
:
:BrCH₂CH₂C₆H₅
:MFCD00000240
:185.06 g/mol
:220…221 °C (760 mmHg)
:–56 °C
:1.35 g/cm³
:89 °C
:203-130-8
:GHS07
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5553 to 1.5573 (20°C, 589 nm)
- Water:≤0.05 %
- GC:≥97.5 %
Frequently Bought Together