Safety gas burners, JUMBO, with stopcock
Supplier: BOCHEM
Certificates
About this item
Safety gas burner JUMBO with stopcock for natural gas/propane.
- Thermo safety element
- Bimetal-flame antena magnet breech and pilot flame
- Air regulation
- Natural gas and propane gas/butane gas
- Max. 1300 °C
Frequently Bought Together