Safety gas burners, with needle valve
Supplier: BOCHEM
Certificates
About this item
Safety gas burner with needle valve and anti-slip rubber coating under the base.
- Thermo safety element
- Pilot flame
- Air regulation
- Max. 1300 °C
Frequently Bought Together