
(±)-2-Phenyl-3-butyn-2-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-66-2
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00004454
:146.19 g/mol
Boiling Pt:92…93 °C (10 mmHg)
Melting Pt:49…53 °C
Density:1.031
Flash Pt:96 °C (205 °F)
EINECS:204-855-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together