Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Hydroxyphenylazo)benzoic acid, Supelco®
2-(4-Hydroxyphenylazo)benzoic acid, Supelco®
Catalog # 54793-1G-F
Supplier:  Merck
CAS Number:  
undefined
2-(4-Hydroxyphenylazo)benzoic acid, Supelco®
Catalog # 54793-1G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.23 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:216-655-2
CAS Number:1634-82-8
Formula:C₁₃H₁₀N₂O₃
MDL Number:MFCD00002428