
5-Bromo-2-(trifluoromethoxy)benzaldehyde 98%
Catalog # H31751.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
CAS Number:923281-52-1
:
Formula:C₈H₄BrF₃O₂
MDL Number:MFCD03701029
:269.02 g/mol
:GHS07
Storage Temperature:Ambient