Order Entry
Czech Republic
ContactUsLinkComponent
Dimethylcyclopropane-1,1-dicarboxylate, Sigma-Aldrich®
Dimethylcyclopropane-1,1-dicarboxylate, Sigma-Aldrich®
Catalog # SIAL412783-50ML
Supplier:  Merck
CAS Number:  
Dimethylcyclopropane-1,1-dicarboxylate, Sigma-Aldrich®
Catalog # SIAL412783-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :158.15 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:414-240-2
CAS Number:6914-71-2
Formula:C₇H₁₀O₄
MDL Number:MFCD00192079