Order Entry
Czech Republic
ContactUsLinkComponent
 
DL-α-Tocopherol acetate, Sigma-Aldrich®
Catalog # SIAL29992-50G-F
Supplier:  Merck
CAS Number:  
DL-α-Tocopherol acetate, Sigma-Aldrich®
Catalog # SIAL29992-50G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :472.75 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:231-710-0
CAS Number:7695-91-2
 :52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042