Order Entry
Czech Republic
ContactUsLinkComponent
DL-α-Tocopherol acetate
DL-α-Tocopherol acetate
Catalog # 47786.
Supplier:  Merck
CAS Number:  
undefined
DL-α-Tocopherol acetate
Catalog # 47786.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Pk Info:
    100 mg

Specifications

About this item

 :
 :472.75 g/mol
EINECS:231-710-0
CAS Number:7695-91-2
 :52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042