Ethyl indole-2-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3770-50-1
:
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00005609
:189.21 g/mol
Melting Pt:119…125 °C
Storage Temperature:Refrigerator
EINECS:223-206-4
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:119 to 125 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together