

1-Bromo-3-methyl-2-butene ≥90% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:870-63-3
:
Formula:C₅H₉Br
MDL Number:MFCD00000242
Boiling Pt:135 °C
Density:1.281
Flash Pt:32 °C (90 °F)
Storage Temperature:Refrigerator
ADR:8,II
:Stabilised with silver
:149.03 g/mol
EINECS:212-799-5
UN:2920
:GHS05
:GHS02
Related Information
Frequently Bought Together