Sulphamide, Sigma-Aldrich®
Catalog # SIAL86033-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:96.11 g/mol
:M-Clarity™ quality level = MQ100
EINECS:232-262-9
CAS Number:7803-58-9
Formula:H₄N₂O₂S
MDL Number:MFCD00011606