1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:174899-82-2
:
Formula:C₈H₁₁F₆N₃O₄S₂
MDL Number:MFCD03788927
:391.32 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together