
Diglycolic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-99-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00004309
:134.09 g/mol
Melting Pt:141…145 °C
Storage Temperature:Ambient
EINECS:203-823-5
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:141 to 145 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together