1-Hexyl-3-methylimidazolium iodide, Sigma-Aldrich®
Catalog # SIAL713058-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:294.18 g/mol
CAS Number:178631-05-5
Formula:C₁₀H₁₉IN₂
MDL Number:MFCD12964976