Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluorosalicylaldehyde
5-Fluorosalicylaldehyde, Sigma-Aldrich®
5-Fluorosalicylaldehyde, Sigma-Aldrich®
SIAL526274-1G
:
Merck
:
5-Fluorosalicylaldehyde, Sigma-Aldrich®
SIAL526274-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
140.11 g/mol
:
M-Clarity™ quality level = MQ100
:
347-54-6
:
C₇H₅FO₂
:
MFCD01090997