4-Chlorobutyryl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4635-59-0
:
Formula:C₄H₆Cl₂O
MDL Number:MFCD00000754
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:225-059-1
UN:2927
Frequently Bought Together
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Ethylacetát ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Petrolether, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerin 99.6% ACS
Supplier: Thermo Fisher Scientific
3-Chloropropionyl chloride, Sigma-Aldrich®
Supplier: Merck
Chloroform ≥99.9% stabilizovaný, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Etylenglykol, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Isooktán ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-methyl-2-pyrolidon ≥99.5%, Biograde for DNA and peptides synthesis
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerin solution 86-89% (by titrimetric analysis), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals