
4-Bromo-1,3-dimethoxybenzene, Sigma-Aldrich®
Catalog # SIAL157554-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:217.06 g/mol
Boiling Pt:282 °C (1013 hPa)
Density:1,507 g/cm³ (20 °C)
EINECS:241-717-0
:GHS07
CAS Number:17715-69-4
Formula:C₈H₉BrO₂
MDL Number:MFCD00009844