
1-Bromodecane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-29-8
:
Formula:BrH₂C(CH₂)₈CH₃
MDL Number:MFCD00000221
:221.18 g/mol
Boiling Pt:238 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:203-955-3
UN:2810
ADR:6.1,III
:GHS09
Frequently Bought Together