Order Entry
Czech Republic
ContactUsLinkComponent
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3007300
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3007300
Catalog # REGE3007300
Supplier:  REAGECON
CAS Number:  
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 3007300
Catalog # REGE3007300
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499