Organic reference standard, Picric acid 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17890500AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:88-89-1
- Description:Picric acid 100 µg/ml in acetonitrile
- Formula:C₆H₃N₃O₇
- Description:Picric acid