Order Entry
Czech Republic
ContactUsLinkComponent
Cobalt(II) meso-tetraphenylporphine ≥85% (dye content), Sigma-Aldrich®
Cobalt(II) meso-tetraphenylporphine ≥85% (dye content), Sigma-Aldrich®
Catalog # SIAL252190-500MG
Supplier:  Merck
CAS Number:  
Cobalt(II) meso-tetraphenylporphine ≥85% (dye content), Sigma-Aldrich®
Catalog # SIAL252190-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :671.65 g/mol
Density:1,276 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:238-018-8
CAS Number:14172-90-8
Formula:C₄₄H₂₈CoN₄
MDL Number:MFCD00010725