4-Bromobutyronitrile, Sigma-Aldrich®
: Merck
:5332-06-9
:
:BrH₂CCH₂CH₂CN
:MFCD00001971
:148 g/mol
:205 °C (1013 hPa)
:1.49 g/cm³ (20 °C)
:103 °C
:226-236-6
:6.1,III
:M-Clarity™ quality level = MQ200
:3276
Frequently Bought Together