
2-Acetylthiophene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-15-3
:
Formula:C₆H₆OS
MDL Number:MFCD00005442
:126.18 g/mol
Boiling Pt:214 °C (1013 hPa)
Melting Pt:9 °C
Density:1.1704 g/cm³ (20 °C)
EINECS:201-804-6
UN:2810
ADR:6.1,II
:GHS06
Frequently Bought Together