Ethyl oleate, Sigma-Aldrich®
Catalog # SIAL317675-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
:
:310.52 g/mol
Merck Index:12,06965
Boiling Pt:386 °C (1013 hPa)
Melting Pt:–32 °C
Density:0.87 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:174 °C
Storage Temperature:Ambient
EINECS:203-889-5
CAS Number:111-62-6
:85049-36-1
Formula:C₂₀H₃₈O₂
MDL Number:MFCD00009579