Manganese ≥99.9998% (metals basis), pieces ≤ 10 mm, Ultrapure
Catalog # 45130.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:7439-96-5
:
Formula:Mn
MDL Number:MFCD00011111
:54.94 g/mol
Storage Temperature:Ambient
EINECS:231-105-1