
Caffeine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:58-08-2
:
Formula:C₈H₁₀N₄O₂
MDL Number:MFCD00005758
Merck Index:13,01636
Melting Pt:235…239 °C
Density:1.23 g/cm³ (19 °C)
:194.19 g/mol
EINECS:200-362-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together