Urea, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Merck Index:13,09935
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
:60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
Frequently Bought Together