3-Nitrophenol for synthesis, Sigma-Aldrich®
: Merck
:554-84-7
:
:C₆H₅NO₃
:MFCD00007240
:139.11 g/mol
:13,06652
:271 °C (1013 hPa)
:94…95 °C
:1.485 g/cm³ (20 °C)
:Ambient
:209-073-5
:1663
:6.1,III
:M-Clarity™ quality level = MQ200
:>100 °C
:GHS07
- Solubility:(20 °C) insoluble
- Melting Point:95 - 98 °C
- Molar Mass:139.11 g/mol
- Bulk Density:640 kg/m3
- Boiling Point:194 °C (93 hPa)
- Flash Point:100 °C
- Density:1.49 g/cm3 (20 °C)
- pH:6.6 - 8.6 (H2O, 20 °C)
- Ignition Point:400 °C
- Synonyms:m-Nitrophenol
- Hill Formula:C6H5NO3
- Chemical Formula:O2NC6H4OH
Frequently Bought Together