Zadání objednávky
Czech Republic
ContactUsLinkComponent
Beaker tongs, stainless steel, polished
Beaker tongs, stainless steel, polished
Catalog # RSGA043.230
Supplier:  RSG
undefined
Beaker tongs, stainless steel, polished
Catalog # RSGA043.230
Supplier:  RSG
Specifications
  • Délka:
    230 mm
  • Bal.:
    1
  • Upínací oblast:
    15 - 60 mm
Specifications
About this item
These beaker tongs are made from electrolytically polished stainless steel.
  • Jaws with cork inserts