N-Methylaniline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-61-8
:
Formula:C₇H₉N
MDL Number:MFCD00008283
:107.16 g/mol
Merck Index:12,06098
Boiling Pt:195 °C (760 mmHg)
Melting Pt:–57 °C
Density:0.987 g/cm³
Flash Pt:86 °C
Storage Temperature:Ambient
EINECS:202-870-9
UN:2294
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5685 to 1.5725 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.003 %
- Carbonyl compounds:≤0.01 % (as formaldehyde)
- Titrable acid:≤0.001 meq/g
Specification Test Results
Frequently Bought Together