


2,4-Dinitroaniline 99% (dry weight) wetted with ca. 15% water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-02-9
:
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007151
:183.12 g/mol
Merck Index:14,3270
Boiling Pt:401 °C (1013 hPa)
Melting Pt:176…180 °C
Density:1.61
Flash Pt:224 °C (435 °F)
Storage Temperature:Ambient
UN:1596
ADR:6.1,II
:moistured with water
EINECS:202-553-5
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together