

3-Nitroaniline 98%
:99-09-2
:
:C₆H₆N₂O₂
:MFCD00007782
:138.13 g/mol
:12,06679
:306 °C (1013 hPa)
:111…114 °C
:0,9 g/cm³ (25 °C)
:196 °C
:Ambient
:202-729-1
:1661
:6.1,II
:GHS06
:GHS08
- Appearance:Yellow to ochre to orange Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:111 to 114 °C
- GC:≥97.5 %
Frequently Bought Together