ContactUsLinkComponent
Filter plates, 96-well, GST MultiTrap™
Filter plates, 96-well, GST MultiTrap™
Catalog # 28-4055-00
Supplier:  Cytiva
undefined
Filter plates, 96-well, GST MultiTrap™
Catalog # 28-4055-00
Supplier:  Cytiva
Specifications
  • Application:
    Protein purification
  • No. of wells:
    96
  • Description:
    GST MultiTrap™ 4B
  • Material:
    PP/PE
  • Pk:
    4
  • Well volume:
    800 µl
Specifications
About this item
GST MultiTrap™ FF and GST MultiTrap™ 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and purification of glutathione S-transferase (GST) tagged proteins.
  • Highly reproducible well-to well and plate-to-plate results
  • High purity in one step, high chemical stability
  • Load unclarified samples directly to the wells
  • Wells are prepacked with Glutathione Sepharose™ media for convenience
Purification of up to 0,5 mg of GST-tagged proteins/well directly from unclarified cell lysate can be achieved using GST MultiTrap™ filter plates. Direct purification of GST-tagged proteins from unclarified lysate shortens handling time and minimizes degradation of sensitive target proteins. The 96-well plate format gives great flexibility, both when working with automated robotic systems and manually using centrifugation or vacuum. Consistent well-to-well and plate-to-plate performance ensures high reproducibility.
Delivery: GST MultiTrap™ FF is prepacked with Glutathione Sepharose™ 4 Fast Flow, while GST MultiTrap™ 4B is prepacked with Glutathione Sepharose™ 4B.