Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-4-nitrophenol
3-Fluoro-4-nitrophenol ≥99%
3-Fluoro-4-nitrophenol ≥99%
3-Fluoro-4-nitrophenol ≥99%
A14350.06
:
Thermo Fisher Scientific
:
3-Fluoro-4-nitrophenol ≥99%
A14350.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
157.1 g/mol
:
91…93 °C
:
206-895-6
:
GHS06
:
GHS07
:
Ambient
:
394-41-2
:
C₆H₄FNO₃
:
MFCD00041251