
3-Fluoro-4-nitrophenol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:394-41-2
:
Formula:C₆H₄FNO₃
MDL Number:MFCD00041251
:157.1 g/mol
Melting Pt:92…95 °C
EINECS:206-895-6
UN:2811
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:91 to 95 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together