Order Entry
Czech Republic
ContactUsLinkComponent
 
 
5-Fluoro-2-nitrophenol 99%
 
undefined
 
5-Fluoro-2-nitrophenol 99%

 

  • Appearance:
    Yellow to orange-brown Powder or crystals or crystalline mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    32 to 35 °C
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together