Order Entry
Czech Republic
ContactUsLinkComponent
2-(Trifluoromethoxy)benzoic acid ≥98%
Irritant
2-(Trifluoromethoxy)benzoic acid ≥98%
Catalog # ABCRAB119307.0001
Supplier:  abcr
CAS Number:  
Irritant
2-(Trifluoromethoxy)benzoic acid ≥98%
Catalog # ABCRAB119307.0001
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.12 g/mol
Melting Pt:78…80 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1979-29-9
Formula:C₈H₅F₃O₃
MDL Number:MFCD00052325